Lead(II) bromide

Supplier: TCI America
Danger

Synonyms: Lead dibromide

L0288-1G L0288-25G L0288-5G
TCL0288-1GEA 45.09 CAD
TCL0288-1G TCL0288-25G TCL0288-5G
Lead(II) bromide
Lead (II) bromide
Formula: Br₂Pb
MW: 367.01 g/mol
Boiling Pt: 916
Melting Pt: 362 (lit.)
MDL Number: MFCD00011156
CAS Number: 10031-22-8
EINECS: 233-084-4
UN: 2291
ADR: 6.1 III
Merck Index: 14,05403
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0df59e07414b9a39a79fae159c6201a8e5d8fd7a&vid=0df59e07414b9a39a79fae159c6201a8e5d8fd7a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR