XLD Agar, Dehydrated Culture Media, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
MH031-2.5KG MH031-5KG
CA89222-274EA 3841.22 CAD
CA89222-274 CA89222-276
XLD Agar, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Xylose-Lysine Deoxycholate Agar (XLD Agar) is a selective medium recommended for the isolation and enumeration of Salmonella Typhi and other Salmonella species from clinical and non clinical samples.
Order Now
SPECIFICATIONS
Environmentally Preferable