Lead(II) iodide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
Lead diiodide
L0279-1G
L0279-25G
L0279-5G
TCL0279-1GEA
36.34
CAD
TCL0279-1G
TCL0279-25G
TCL0279-5G
Lead(II) iodide ≥98.0% (by titrimetric analysis)
Lead (II) iodide
Formula:
I₂Pb Boiling Pt: 954 °C (lit.) Melting Pt: 412 °C (lit.) |
MDL Number:
MFCD00011163 CAS Number: 10101-63-0 EINECS: 233-256-9 UN: 2291 ADR: 6.1,III Merck Index: 14,05411 |
|
Specification Test Results
Purity (Chelometric Titration) | Min. 98.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...