SureVector, Vectors, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
G7514A G7516A G7516B
CA76193-662EA 653.37 CAD
CA76193-662 CA76193-664 CA76193-666 CA76193-668 CA76193-670 CA76193-672 CA76193-674 CA76193-676 CA76193-678 CA76193-680 CA76193-682 CA76193-684
SureVector, Vectors, Agilent Technologies
Vectors, Plasmids and Libraries
Combine standard DNA components to build custom vector constructs from Agilent’s set of standard parts.
  • Rapid custom vector generation - Less than a day from design to vector, compared to 4 weeks for custom vector services
  • Reliable and precise assembly - SureVector is extensively validated to ensure that standard parts can be interchanged, without loss of functionality
  • More flexible than traditional systems - Assemble new vectors in your lab as experimental requirements change, versus always ordering a new one
  • Control your experiments - Take control of your experiments, troubleshoot your own work, not that of your service providers DNA assembly
SureVector next-generation cloning uses the power of synthetic biology to combine standard DNA components to build custom vector constructs from the Agilent set of standard parts. Simply design a vector, order or PCR your gene-of-interest, and your custom plasmid is just a single reaction away.
Proprietary enzymes enable rapid and reliable assembly of multiple DNA fragments into a circular, recombinant plasmid containing your target gene.
Delivery information: Components include: bacterial selection markers, bacterial origins of replication, mammalian selection markers, promoters for yeast, mammalian cells and bacteria, and a variety of solubility/affinity tags; Enabling functionality in a range of cells.
Caution: For research use only. Not for use in diagnostic procedures.