GeneJammer, Transfection Reagents, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
204131 204130 204132
CA99900-708EA 1631.5 CAD
CA99900-708 CAST204130 CAST204132
GeneJammer, Transfection Reagents, Agilent Technologies
Transfection Reagents
The GeneJammer transfection reagent is a proprietary formulation of novel polyamines ideal for transient or stable transfections in a wide variety of cell types.
  • Fast and easy to use, no media changes required
  • Enables high level adenovirus transduction in the absence of an adenovirus specific receptor
  • Works with media containing antibiotics, growth factors and/or other supplements
This transfection reagent is effective in both serum-containing medium and serum-free medium, offering high efficiencies with minimal cytotoxicity.
1.33 mg/ml reagent in 80% ethanol.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable