AriaMx Realtime PCR System, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
401427 401490 401490 401491 401492 401493 401494 5190-7702 5190-7708 5190-9370 600906 600907 G8830-67004 401425 401428
99901-796EA 42.06 CAD
99901-796 76193-374 CA76193-374 CA76193-376 CA76193-584 CA76193-632 CA76193-634 CA76193-638 CA76193-640 CA76193-644 CA76193-692 CA76193-694 CA76193-696 CA76193-698 CA76193-724 CA76193-726 CA76193-728 CA76193-730 CA76193-732 CA76208-058 CA76208-060 CAST401425 CAST401428
AriaMx Realtime PCR System, Agilent Technologies
Thermal Cyclers
The AriaMx Real-time PCR System is a fully integrated quantitative PCR amplification, detection, and data analysis system.
  • Gene expression analysis
  • New genotyping/HRM capability, mRNA quantification
  • NGS quantification: library preparation, result validation, nucleic acid monitoring, rare allele detection
  • Can use self-Installable optical cartridges
  • Operates at a Cq uniformity standard deviation of <0.20 at fast cycling (5 sec at 95 °C / 10 sec at 60 °C)
  • Data acquisition time of <3 sec/plate
  • Dynamic range of 9 orders of magnitude
  • Operates at a maximum
More Product Information