AdEasy Adenoviral Vector Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
240006
240005
240081
240082
240007
240008
240009
240010
240085
CA95040-764EA
556.82
CAD
CA95040-764
CA95040-766
CA97066-368
CA97066-370
CA99900-934
CA99900-936
CA99900-938
CA99900-940
CAST240085
AdEasy Adenoviral Vector Systems, Agilent Technologies
Vectors, Plasmids and Libraries
The AdEasy system saves you a month of work over traditional methods by producing the recombinant adenoviral plasmid by homologous recombination in E. coli. The AdEasy XL system includes BJ5183 cells pre-transformed with the pAdEasy-1 plasmid, this feature dramatically decreases background caused by the non-recombinant shuttle plasmid.
- 300% more recombinants with specialty competent cells
- Adherent AD-293 cells improve viral packaging
- Infects dividing and non-dividing cells
Caution: For Research Use Only. Not for Use in Diagnostic Procedures.