AAV Helper-Free System, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
240074 240075
CA97066-364EA 607.55 CAD
CA97066-364 CA97066-366
AAV Helper-Free System, Agilent Technologies
Vectors, Plasmids and Libraries
The AAV Helper-Free System eliminates the requirement for wild-type adenovirus co-infection from both AAV vector production and AAV stock titering steps, making this system entirely helper virus-free and safer. This system works with a broad range of hosts and infects both dividing and non-dividing cells.
  • Stable gene expression
  • High titer virus
  • Lower toxicity vs. non-viral methods of gene delivery
Caution: For Research Use Only. Not for Use in Diagnostic Procedures.