Synthetic Sea Salt, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M1344-500G
M1344-2.5KG
M1344-5KG
CA95020-976EA
1402.96
CAD
CA95020-976
CA89223-780
CA89223-782
Synthetic Sea Salt, HiMedia Laboratories
Media for Microbiology
Additives and Supplements for Microbiology
Synthetic sea salt is recommended for preparation of special diluents.
- Stored at 2 to 8 °C (prepared medium)
It is used as a diluent during detection and enumeration of E. coli and coliforms in surface and wastewater. Sodium chloride in the medium helps to maintain osmotic
balance and the other ingredients provide buffering action.