Media Bottles, PETG, WHEATON®
Supplier: DWK Life Sciences (KIMBLE)
WPBGC2000S WPBGC1000S WPBGC0500S WPBGC0250S WPBGC0125S WPBGC0060S WPBGC0030S WPBGC2000SB WPBGC1000SB WPBGC0250SB
10112-168CS 246.56 CAD
10112-168 10112-170 10112-172 10112-174 10112-176 10112-178 10112-180 10112-182 10112-184 10112-186
Media Bottles, PETG, WHEATON®
Bottles Cell Culture Bottles
These bottles are made of eastman tritan eastar copolyester PETG resin for single use labware and medical devices.
  • Ideal for the storage of media, laboratory reagents and biological buffers
  • Graduation markings are molded in for reference filling
  • Have a standard cap
  • Comprised of resins that are free of animal-derived components
  • Suitable for transport and storage, leak resistant
  • Recommended temperature range +70 to –40 °C
The PETG is an amorphous material that displays performance properties including clarity and toughness as well as excellent gasMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable