Macro Cuvettes with Screw Cap, Type 41, Fireflysci
Supplier: FIREFLYSCI, INC.
41UV1 41UV2 41UV5 41UV10 41UV50 41UV100
76016-058EA 1339.38 CAD
76016-058 76016-060 76016-062 76016-064 76016-066 76016-068
Macro Cuvettes with Screw Cap, Type 41, Fireflysci
Cuvettes Spectrophotometer Cuvettes
The Type 41 cuvette is standard screw cap cell which perform anaerobic work.
  • Fire-fused to ensure that cuvettes are robust and durable
  • Two windows optically polished
  • Features 13-425 screw cap threadings for maximum durability and versatility
  • Rated to handle up to 5 bar of pressure
The open SEPTA screw cap enables researchers to make sterile injections into the cuvette without compromising the experiment.
Delivery information: Includes one SEPTA screw cap and one closed screw cap.