GENIE Temp-Shaker 100™, Scientific Industries
Supplier: Scientific Industries
SI-G100
76226-090EA 5813.8 CAD
76226-090
GENIE Temp-Shaker 100™, Scientific Industries
Shakers Shaking Incubators
Gentle Orbital Shaking from 20 to 100 rpm, while providing precise temperature control from 28 to 75 °C, for use in many laboratory applications.
  • Unique 3-way access clear dome chamber provides 360° view at all times
  • Quick access to your samples with easy tilt-n-rest feature using the exclusive "kick stand" feature to open the chamber cover, left, right, or back, to optimize bench space
  • The lightweight cover can also be easily removed for cleaning and attaching accessories
  • Unique airflow design can heat the chamber to a stable 75 °C in as little as 20 minutes
  • With the chamber door open, unit will maintain the chamber temperature within ±3 °C
Various voltages and plugs available for operation around the world. Product is produced and shipped from within the US.
Accessories information: No additional accessories necessary for operation of this model. However, stacking tray is available to double capacity. No tools necessary to mount accessories.
Delivery information: This model includes the non-slip tray with drip-lip to contain any spills. Non-slip stacking tray and additional interchangeable platform styles available, sold as accessories.