Backyard Brains® Neuron Spikerbox
Supplier: BACKYARD BRAINS, INC.
BYBP0005 BYBP0013 BYBP0105 BYBP0107
470343-158EA 18.4 CAD
470343-158 470343-164 470353-928 470353-926
Backyard Brains® Neuron Spikerbox
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Experiment with the nervous system.
  • Free software download
  • Classic and Pro versions
  • The easiest way to visualize and record neurological data
Record living neurons, hear what the brain sounds like, and see action potentials on your smartphone or PC. Never before have powerful neuroscience tools been this accessible.
The classic is the model you have used for years, powerful and functional, ideal for basics. Perfect for science fairs, you can track, measure, and experiment. Just supply invertebrates (cricket, cockroach, earthworm) and a hypothesis. The Pro model adds additional inputs, improved sensitivity, a more resilient case, and improved speaker and electronics. Both kits are easy to use for beginners, but powerful enough to record publication-ready results. This kit can turn any home or classroom into a Ph.D. lab!