DAVISIL® Small Pore Chromatographic Silica, W.R. Grace
Supplier: W.R. GRACE & CO. - CONN
5173177
5139180
76210-932EA
26668.39
CAD
76210-932
76210-934
DAVISIL® Small Pore Chromatographic Silica, W.R. Grace
Chromatography Sorbents
HPLC Sorbents
Proven performance and high quality chromatographic silica.
- Predicable scale-up
- Exceptional quality and consistency
- High capacity
Grace Davisil® 23 Å and 35 Å Chromatographic Silica is suitable for use as a packing for liquid chromatography normal phase separations. Applications include separation and purification of small molecule pharmaceuticals and organic synthesis. High surface area boosts capacity. Scalable from analytical to process scale.