Vydac® Chromatographic Silica, W.R. Grace
Supplier: W.R. GRACE & CO. - CONN
Vydac® chromatographic resin is excellent for biopurification. Proven in applications for protein, peptide, and oligonucliotide separations.
- Completely scalable to ton quantities
- High recovery of peptides and proteins
- Available in a wide range of bonding chemistries
Grace pioneered one of the first platforms for reverse phase protein and peptide separation. Referenced in more than 9000 patents, Vydac® resin is the phase to which all others are compared. It is relied upon in numerous commercial scale protein purifications. High recovery of proteins makes this product more cost effetive than alternatives and is available in a wide range of bonding chemistries for alternate selectivities.