Arduino Uno Development Board
Supplier: NEWARK ELECTRONICS
78T1601
470237-736EA 45.69 CAD
470237-736
Arduino Uno Development Board
Educational Materials Physics Educational Materials Applied Physics Learning Activities
The Arduino Uno is a microcontroller board based on the ATmega328.
  • Used for communications and networking
  • Perfect for advanced programming use
  • Microcontroller Atmel ATmega328
  • Operating voltage of 5 V
  • Maximum supply voltage 6 to 20 V
  • Digital I/O pins 14 (of which 6 provide PWM output)
  • DC current per I/O pin 40 mA
  • DC current for 3.3 V pin 50 mA
  • Flash memory 32 KB of which 0.5 KB is used by boot loader
  • SRAM 2 KB, EEPROM 1KB
  • Clock speed 16 MHz
  • Can be powered via the USB connection or with an external power supply
This board has 14 digital input/output pins (of which 6 can be used as PWM outputs), 6 analog inputs, a 16 MHz ceramic resonator, a USB connection, a power jack, an ICSP header, and a reset button. It contains everything needed to support the microcontroller; simply connect it to a computer with a USB cable or power it with a AC-to-DC adapter or battery to get started. The Uno differs from all preceding boards in that it does not use the FTDI USB-to-serial driver chip. Instead, it features the Atmega16U2 (Atmega8U2 up to version R2) programmed as a USB-to-serial converter.