Arduino Micro Development Board
Supplier: NEWARK ELECTRONICS
63W3544
470237-730EA 67.19 CAD
470237-730
Arduino Micro Development Board
Educational Materials Physics Educational Materials Applied Physics Learning Activities
An Arduino Micro development board based on the ATmega32u4 microcontroller.
  • Used for embedded design and development
  • Perfect for advanced programming use
  • 6MHz crystal oscillator
  • ICSP header
  • Reset button
  • Micro USB connection
  • 20 digital input/output pins (of which 7 can be used as PWM outputs and 12 as analog inputs)
  • Input voltage (recommended) range from 7 to 12 V
  • Input Voltage (limit) range from 6 to 20 V
  • 32 KB flash memory of which 4KB used by bootloader
  • 2.5 KB SRAM
  • 1 KB EEPROM
The ATmega32u4 is a 8bit AVR RISC based microcontroller featuring 32 KB flash memory, 2.5 KB SRAM, 1 KB EEPROM, USB 2.0 full speed/low speed device, 10bit A to D converter and JTAG interface for on chip debug. The board contains everything needed to support the microcontroller, simply connect it to a computer with a micro USB cable to get started. It has a form factor that enables it to be easily placed on a breadboard. The Micro is similar to the Arduino Leonardo in that the ATmega32u4 has built in USB communication, eliminating the need for a secondary processor. This allows the Micro to appear to a connected computer as a mouse and keyboard, in addition to a virtual (CDC) serial/COM port. It also has other implications for the behaviour of the board. The Arduino Micro can be powered via the micro USB connection or with an external power supply. The power source is selected automatically.