Quintix® Balances, Standard Models, 100 - 240 V, Sartorius
Supplier: Sartorius
QUINTIX65-1S QUINTIX125D-1S
CA101485-988EA 3783.84 CAD
CA101485-988 CA101485-990 CA101485-992 CA101485-994 CA101485-996 CA101485-998 CA101486-000 CA101486-002 CA101486-004 CA101486-030 CA101486-032 CA101486-034 CA102355-766 CA102355-768 CA102355-770 CA102355-772 CA102355-774 CA102355-776 CA102355-778 CA490019-406 CA490019-408
Quintix® Balances, Standard Models, 100 - 240 V, Sartorius
Balances and Scales
Quintix® balances are designed with the combination of intuitive touch-screen and most precise weighing technology to achieve the best results. The enhancement of user interface with built-in application programs and color touch screen helps in intuitive operation of the balances.
  • Direct data transfer and ergonomic draft shield
  • Chemical resistant finish at the top housing
  • Integrated application programs and intuitive user guidance for greater efficiency
These balances are features with the isoCAL fully automatic internal adjustment to ensure highest accuracy and convenience.
Delivery information: Supplied with universal power adapter, in-use cover, application guide and dust cover for analytical balances.