Straight-Sided Wide-Mouth Jars, HDPE, Environmental Express®
Supplier: Environmental Express
BPC1120 BPC1172
89202-832CS 57.89 CAD
89202-832 89202-844
Straight-Sided Wide-Mouth Jars, HDPE, Environmental Express®
Jars General Use Jars
Gain easier access to samples.
  • Large diameter lid allows for easier scooping and removal of samples
  • Reach entire sample with straight-sided design
  • Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory
  • Level 3 containers meet EPA guidelines as well, but do not ship with a certificate of analysis
Order Now
SPECIFICATIONS
Environmentally Preferable