Oblong Wide Mouth Bottles, Pre-Cleaned, HDPE, Environmental Express®
Supplier: Environmental Express
BPC1505 BPC1501 BPC1502
89202-824CS 58.78 CAD
89202-824 89202-828 89202-830
Oblong Wide Mouth Bottles, Pre-Cleaned, HDPE, Environmental Express®
Bottles Plastic Bottles
Store and retrieve samples with ease.
  • Oblong shape minimizes dead space between bottles when stored
  • Keep samples safe with leakproof cap
Level 1 containers are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory.
Level 3 containers meet EPA guidelines as well, but do not ship with a certificate.
Order Now
SPECIFICATIONS
Environmentally Preferable