Energy Saving Forced Convection Ovens, Yamato
Supplier: Yamato Scientific
DNE-411 DNE-611 DNE-811 DNE-911
76209-638EA 8500.13 CAD
76209-638 76209-640 76209-642 76209-644
Energy Saving Forced Convection Ovens, Yamato
Ovens Standard Ovens
Energy saving, programmable forced convection eco-oven with lower power consumption and CO2 emission.
  • High precision controller allows high performance temperature control and display of CO2 and power discharge
  • Maximum temperature reaching time reduced by 15 minutes (no-load) compared to previous models
  • Program operation with a maximum of 99 steps, 99 patterns repeatable
  • Easy system upgrade with various option settings
Heat tightness and insulation design of the chamber achieves an energy saving rate of about 40% at constant temperature compared to previous models.
Users can adjust the temperature setting with a digital key. Operation functions include fixed-temperature operation, program operation, and duration/timer operation functions. Display functions include integration time, calendar time, calibration offset, power consumption, CO2 discharge, heater operation, and time display. Safety features such as self-diagnostic functions, a key lock function, and an electric leakage breaker with over-current protection are provided.
A TRIAC zero-cross control manages the heater circuit. Ovens feature one Scirocco fan, and the largest unit features two. They also feature a stainless pipe heater, and the largest two units have two each. The ovens' interior is made of stainless steel, and the exterior features electro-galvanized steel sheet with epoxy and melamine-resin baking finish. Brackets and a shelf are provided.