Cole-Parmer® Freezer/Mill® Cryogenic Grinders
Supplier: SPEX
6775-115 6775-230 6875-115 6875-230 6875A-115 6875A-230 6875D-115 6875D-230
76181-976EA 24380.33 CAD
76181-976 76181-978 76182-004 76182-006 76182-008 76182-010 76182-012 76182-014
Cole-Parmer® Freezer/Mill® Cryogenic Grinders
Mills
Grind the toughest samples at cryogenic temperatures.
  • Reach cryogenic temperatures in minutes
  • Dual grinding and cooling chambers available
  • User-friendly touch screen controls
  • 20 user-defined programs for easy quick recall
  • Enhanced safety features
The Freezer/Mill® programmable cryogenic grinder pulverizes through the toughest samples at cryogenic temperatures. Samples are placed in a vial and immersed in a self-contained insulated liquid nitrogen (LN2) tub, reaching cryogenic temperatures in minutes, where they are then pulverized with a magneticallyMore Product Information