European Pharmacopeia Arsenic Limit Test, Ace Glass
Supplier: Ace Glass
6301-01 6301-02 6301-10
76214-392EA 553.19 CAD
76214-392 76214-394 76214-396
European Pharmacopeia Arsenic Limit Test, Ace Glass
Distillation Components Distillation Sets
Arsenic Limit Test apparatus as described in the European Pharmacopeia chapter 2.4.3.
Complete apparatus consists of a 125 mL erlenmyer flask, a 24/40 chimney tube with a flat ground O-ring joint and 1 mm drip tip, a test tube, and springs for securing the chimney tube to the bent tube.