Condensers for Giant Soxhlet Extraction Apparatus, Ace Glass
Supplier: Ace Glass
6810-05 6810-15 6810-25
76214-414EA 949.91 CAD
76214-414 76214-416 76214-418
Condensers for Giant Soxhlet Extraction Apparatus, Ace Glass
Condensers
Bulb-type condenser for use with Giant Soxhlet Extraction Apparatus.
  • High-quality borosilicate glass
  • Has Ace-Thred inlet/outlet
  • Standard taper inner bottom joint
  • Tube-end top