Small Capacity Glas-Col® Stirmantle, Ace Glass
Supplier: Ace Glass
12046-01 12046-02 12046-10 12046-13 12046-15 12046-17 12046-20 12046-23 12046-25 12046-27
76214-474EA 2783.95 CAD
76214-474 76214-476 76214-478 76214-480 76214-482 76214-484 76214-486 76214-488 76214-490 76214-492
Small Capacity Glas-Col® Stirmantle, Ace Glass
Heaters Heating Mantles
Mantle stirring system with coil induced Synchostart magnetic field, exclusive to Glas-Col.
  • Variable speed StirControl II controller has a 4 foot cord and can be placed outside the hood
  • StirControl II features two receptacles for operating two Stirmantles at once
  • Complete system includes mantle, StirControl II, PTFE stirbar, and cord
Stirmantle with cords only does not include StirControl II or PTFE stirbar, for heating needs.
Ordering information: 230 V products are not returnable.