Giant Extractor Flasks with Side Neck, Ace Glass
Supplier: Ace Glass
6885-33 6885-34 6885-35
76214-452EA 292.32 CAD
76214-452 76214-454 76214-456
Giant Extractor Flasks with Side Neck, Ace Glass
Flasks Round Bottom Flasks
Round bottom flasks designed for use with ACE giant extractor apparatus.
  • High-quality borosilicate glass
  • Available in 1000 mL, 3000 mL or 12000 mL capacities
The 24/40 side neck for sampling and/or temperature measurement.