Mouse Recombinant Platelet-Derived Growth Factor Receptor alpha
Supplier: Bon Opus Biosciences
C658-50UG C658-500UG C658-1MG C658-10UG
103529-266EA 372.88 CAD
103529-266 103529-340 103529-398 103529-416
Mouse Recombinant Platelet-Derived Growth Factor Receptor alpha
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Platelet-derived Growth Factor Receptor Alpha is produced by our Mammalian expression system and the target gene encoding Gln24-Glu524 is expressed with a 6His at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.