Human Galnt7 Cd50-1Mg, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CX50-1MG CX50-500UG CX50-50UG CX50-10UG
102498-044EA 3981.6 CAD
102498-044 102498-046 102498-048 102509-354
Human Galnt7 Cd50-1Mg, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human N-acetylgalactosaminyltransferase 7 is produced by our Mammalian expression system and the target gene encoding Pro30-Val657 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.