Recombinant Interleukin-21 receptor
Supplier: Bon Opus Biosciences
CS91-10UG CS91-500UG CS91-1MG CS91-50UG
103529-196EA 83.74 CAD
103529-196 103529-312 103529-386 103529-470
Recombinant Interleukin-21 receptor
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Interleukin-21 Receptor is produced by our Mammalian expression system and the target gene encoding Cys20-Pro236 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.