Recombinant Protein S100-A11
Supplier: Bon Opus Biosciences
CR31-10UG CR31-1MG CR31-50UG CR31-500UG
103529-184EA 83.74 CAD
103529-184 103529-374 103529-458 103529-500
Recombinant Protein S100-A11
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse S100A11 is produced by our E.coli expression system and the target gene encoding Met1-Ile98 is expressed with a 6His at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.