Mouse Recombinant Carbonic anhydrase 12
Supplier: Bon Opus Biosciences
CK69-10UG CK69-50UG CK69-500UG CK69-1MG
102970-196EA 249.64 CAD
102970-196 102970-322 102970-416 102970-510
Mouse Recombinant Carbonic anhydrase 12
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Carbonic Anhydrase 12 is produced by our Mammalian expression system and the target gene encoding Ala25-Ser301 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.