Mouse Recombinant Tyrosine-protein kinase receptor TYRO3
Supplier: Bon Opus Biosciences
CK68-10UG CK68-50UG CK68-500UG CK68-1MG
102970-194EA 50.56 CAD
102970-194 102970-320 102970-414 102970-508
Mouse Recombinant Tyrosine-protein kinase receptor TYRO3
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Tyrosine Protein Kinase Receptor TYRO3 is produced by our Mammalian expression system and the target gene encoding Ala31-Ser418 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.