Mouse Recombinant DAND1
Supplier: Bon Opus Biosciences
CK66-10UG CK66-50UG CK66-500UG CK66-1MG
102970-190EA 158 CAD
102970-190 102970-316 102970-410 102970-504
Mouse Recombinant DAND1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse NBL1 is produced by our Mammalian expression system and the target gene encoding Ala17-Asp178 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.