Mouse Recombinant Granulocyte-Macrophage Colony-Stim, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CK02-10UG CK02-50UG CK02-500UG CK02-1MG
102942-282EA 249.64 CAD
102942-282 102942-284 102942-286 102942-288
Mouse Recombinant Granulocyte-Macrophage Colony-Stim, Bon Opus Biosciences
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Granulocyte-Macrophage Colony-Stimulating Factor is produced by our E.coli expression system and the target gene encoding Ala18-Lys141 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.