Human Recombinant Metalloproteinase Inhibitor 4
Supplier: Bon Opus Biosciences
CJ60-10UG CJ60-50UG CJ60-500UG CJ60-1MG
102944-658EA 249.64 CAD
102944-658 102944-660 102944-662 102944-664
Human Recombinant Metalloproteinase Inhibitor 4
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Tissue Inhibitor of Metalloproteinases 4 is produced by our Mammalian expression system and the target gene encoding Cys30-Pro224 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.