Mouse Recombinant HLA class II histocompatibility antigen gamma chain
Supplier: Bon Opus Biosciences
CJ51-10UG CJ51-50UG CJ51-500UG CJ51-1MG
102944-586EA 150.1 CAD
102944-586 102944-588 102944-590 102944-592
Mouse Recombinant HLA class II histocompatibility antigen gamma chain
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse HLADG is produced by our Mammalian expression system and the target gene encoding Gln56-Leu215 is expressed with a mFc, 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.