Human Recombinant Tumor necrosis factor ligand superfamily member 4
Supplier: Bon Opus Biosciences
CJ45-10UG CJ45-50UG CJ45-500UG CJ45-1MG
102944-546EA 249.64 CAD
102944-546 102944-548 102944-550 102944-552
Human Recombinant Tumor necrosis factor ligand superfamily member 4
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human OX40 Ligand is produced by our Mammalian expression system and the target gene encoding Gln51-Leu183 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.