Human Recombinant Growth/differentiation factor 8

Supplier: Bon Opus Biosciences

CJ43-10UG CJ43-50UG CJ43-500UG CJ43-1MG
102944-530EA 431.34 CAD
102944-530 102944-532 102944-534 102944-536
Human Recombinant Growth/differentiation factor 8
Proteins and Peptides

Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products


  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels


Recombinant Human/Mouse/Rat Growth Differentiation Factor 8 is produced by our Mammalian expression system and the target gene encoding Lys262-Ser375 is expressed.


Accessories information: Accessories not included.


Delivery information: Shipped Via FedEx


Packaging: Shipped at ambient temperature via FedEx envelope.


Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.

The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.

Order Now

SPECIFICATIONS

Protein/peptide name Growth/differentiation factor 8
Environmentally Preferable
Protein/peptide type Recombinant
Species Human
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR