Mouse Recombinant Regenerating Islet-derived 2
Supplier: Bon Opus Biosciences
CJ37-10UG CJ37-50UG CJ37-500UG CJ37-1MG
102944-490EA 173.8 CAD
102944-490 102944-492 102944-494 102944-496
Mouse Recombinant Regenerating Islet-derived 2
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Islets of Langerhans regenerating protein 2 is produced by our Mammalian expression system and the target gene encoding Gln23-Ala173 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.