Human Recombinant 1-phosphatidylinositol 5-phosphate 4-kinase 2-alpha
Supplier: Bon Opus Biosciences
CJ35-10UG CJ35-50UG CJ35-500UG CJ35-1MG
102944-474EA 249.64 CAD
102944-474 102944-476 102944-478 102944-480
Human Recombinant 1-phosphatidylinositol 5-phosphate 4-kinase 2-alpha
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human PIP4K2A is produced by our Mammalian expression system and the target gene encoding Met1-Thr406 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.