Human Recombinant Tissue-specific extinguisher 1
Supplier: Bon Opus Biosciences
CJ31-10UG CJ31-50UG CJ31-500UG CJ31-1MG
102944-450EA 83.74 CAD
102944-450 102944-452 102944-454 102944-456
Human Recombinant Tissue-specific extinguisher 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human PRKAR1A is produced by our Mammalian expression system and the target gene encoding Glu2-Val381 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.