Human Recombinant IL-22 Binding Protein
Supplier: Bon Opus Biosciences
CJ29-10UG CJ29-50UG CJ29-500UG CJ29-1MG
102944-434EA 173.8 CAD
102944-434 102944-436 102944-438 102944-440
Human Recombinant IL-22 Binding Protein
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human IL-22 Binding Protein is produced by our Mammalian expression system and the target gene encoding Thr22-Pro231 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.