Human Recombinant Platelet Factor 4
Supplier: Bon Opus Biosciences
CJ27-10UG CJ27-50UG CJ27-500UG CJ27-1MG
102944-418EA 173.8 CAD
102944-418 102944-420 102944-422 102944-424
Human Recombinant Platelet Factor 4
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human C-X-C Motif Chemokine 4 is produced by our Mammalian expression system and the target gene encoding Glu32-Ser101 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.