Human Recombinant F9/Coagulation factor IX/Christmas factor/Plasma thromboplastin component/PTC/Coagulation factor IXa light chain/Coagulation factor IXa heavy chain
Supplier: Bon Opus Biosciences
CI95-10UG CI95-50UG CI95-500UG CI95-1MG
102944-170EA 158 CAD
102944-170 102944-172 102944-174 102944-176
Human Recombinant F9/Coagulation factor IX/Christmas factor/Plasma thromboplastin component/PTC/Coagulation factor IXa light chain/Coagulation factor IXa heavy chain
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Coagulation factor IX is produced by our Mammalian expression system and the target gene encoding Thr29-Thr461 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.