Human Recombinant Thymidine kinase
Supplier: Bon Opus Biosciences
CI64-10UG CI64-50UG CI64-500UG CI64-1MG
102943-938EA 249.64 CAD
102943-938 102943-940 102943-942 102943-944
Human Recombinant Thymidine kinase
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Thymidine kinase 1 is produced by our Mammalian expression system and the target gene encoding Met1-Asn234 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.