Human Recombinant EGFR
Supplier: Bon Opus Biosciences
CI61-10UG CI61-50UG CI61-500UG CI61-1MG
102943-914EA 249.64 CAD
102943-914 102943-916 102943-918 102943-920
Human Recombinant EGFR
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human EGFR is produced by our Mammalian expression system and the target gene encoding Leu25-Ser645 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.