Human Recombinant 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 1
Supplier: Bon Opus Biosciences
CI45-10UG CI45-50UG CI45-500UG CI45-1MG
102943-842EA 249.64 CAD
102943-842 102943-844 102943-846 102943-848
Human Recombinant 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human FBPase 1 is produced by our Mammalian expression system and the target gene encoding Ser2-Tyr471 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.