Human Pigr Ci09, Bon Opus Biosciences
Supplier: Bon Opus Biosciences
CI09-10UG CI09-1MG CI09-500UG CI09-50UG
102499-694EA 158 CAD
102499-694 102499-696 102499-698 102499-700
Human Pigr Ci09, Bon Opus Biosciences
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Poly-Ig receptor is produced by our Mammalian expression system and the target gene encoding Lys19-Arg638 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.