Human Recombinant Tumor necrosis factor ligand superfamily member 18
Supplier: Bon Opus Biosciences
CI05-10UG CI05-1MG CI05-500UG CI05-50UG
102499-680EA 173.8 CAD
102499-680 102509-572 102509-574 102509-576
Human Recombinant Tumor necrosis factor ligand superfamily member 18
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human GITR Ligand is produced by our Mammalian expression system and the target gene encoding Glu74-Ser199 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.