Human Recombinant Intelectin-1
Supplier: Bon Opus Biosciences
CH39-10UG CH39-1MG CH39-500UG CH39-50UG
102499-628EA 173.8 CAD
102499-628 102499-630 102499-632 102499-634
Human Recombinant Intelectin-1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Intelectin-1 is produced by our E.coli expression system and the target gene encoding Thr19-Ser298 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.